α-Bromo-2-chlorotoulene 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

A993448-25G A993448-5G
77119-120EA 33.95 USD
77119-120 77119-122
α-Bromo-2-chlorotoulene 98%
α-Bromo-2-chlorotoulene

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MW: 205.48 g/mol
MDL Number: MFCD00000566
CAS Number: 611-17-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR