1,3-Dibromo-2,4-difluorobenzene 97%

Supplier: Ambeed
Warning

A988540-1G A988540-5G A988540-25G
77119-110EA 14.86 USD
77119-110 77119-112 77573-962
1,3-Dibromo-2,4-difluorobenzene 97%
1,3-Dibromo-2,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MW: 271.89 g/mol
MDL Number: MFCD07368980
CAS Number: 219926-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR