PP121 98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

A986976-10MG A986976-50MG A986976-5MG A986976-1MG
77119-104EA 56.23 USD
77119-104 77119-106 77119-108 77614-710
PP121 98%
PP121

Precaución: for Research Use Only

Formula: C₁₇H₁₇N₇
MW: 319.37 g/mol
MDL Number: MFCD12912434
CAS Number: 1092788-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR