SB-415286 98%

Supplier: Ambeed
Warning

A984794-10MG A984794-1MG A984794-50MG
77119-096EA 36.04 USD
77119-096 77119-098 77119-100
SB-415286 98%
SB-415286

Precaución: for Research Use Only

Formula: C₁₆H₁₀ClN₃O₅
MDL Number: MFCD04039789
CAS Number: 264218-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR