Tectoridin 99.5+%

Supplier: Ambeed
Warning

Synonyms: 7-(beta-D-Glucopyranosyloxy)-5-hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, 5-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Tectorigenin 7-O-beta-D-Glucopyranoside

A982374-1MG A982374-25MG A982374-50MG
77119-090EA 5.31 USD
77119-090 77119-092 77119-094
Tectoridin 99.5+%
Tectoridin

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₁₁
MW: 462.41 g/mol
MDL Number: MFCD01662715
CAS Number: 611-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR