Diethylene glycol monovinyl ether 98% + stabilised

Supplier: Ambeed
Warning

Synonyms: Vinyl Carbitol, 2-(2-Vinyloxyethoxy)ethanol, Di(ethylene glycol) vinyl ether, 2-(2-(Vinyloxy)ethoxy)ethanol

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A978235-100G A978235-25G A978235-500G
77119-076EA 13.39 USD
77119-076 77119-078 77119-080
Diethylene glycol monovinyl ether 98% + stabilised
Diethylene glycol monovinyl ether

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00192027
CAS Number: 929-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR