2-Fluoro-m-xylene 98%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoro-1,3-dimethylbenzene, 1-Fluoro-2,6-dimethylbenzene

A975291-25G A975291-5G
77119-072EA 39.26 USD
77119-072 77119-074
2-Fluoro-m-xylene 98%
2-Fluoro-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉F
MW: 124.16 g/mol
MDL Number: MFCD00039215
CAS Number: 443-88-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR