Boc-Gly-Leu-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-glycyl-L-leucine

A969883-250MG A969883-1G
77119-046EA 21.22 USD
77119-046 77686-997
Boc-Gly-Leu-OH 95%
Boc-Gly-Leu-OH

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₅
MDL Number: MFCD00190801
CAS Number: 51871-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR