N-tert-Butylbenzylamine 95%

Supplier: Ambeed
Danger

Synonyms: N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine

A964335-25G A964335-5G
77119-032EA 10.61 USD
77119-032 77119-034
N-tert-Butylbenzylamine 95%
N-tert-Butylbenzylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
MDL Number: MFCD00008798
CAS Number: 3378-72-1
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR