4-Chloro-2,6-dimethoxypyrimidine 98%
Supplier: Ambeed
|
Warning
|
A963559-1G
A963559-250MG
A963559-25G
A963559-5G
A963559-10G
77119-018EA
22.65
USD
77119-018
77119-020
77119-022
77119-024
77686-990
4-Chloro-2,6-dimethoxypyrimidine 98%
4-Chloro-2,6-dimethoxypyrimidine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂O₂ |
MDL Number:
MFCD00006068 CAS Number: 6320-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...