6-Bromo-2-chlorobenzothiazole 97%

Supplier: Ambeed
Warning

A963002-10G A963002-1G A963002-25G A963002-5G
77119-010EA 55.62 USD
77119-010 77119-012 77119-014 77119-016
6-Bromo-2-chlorobenzothiazole 97%
6-Bromo-2-chlorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MDL Number: MFCD04971822
CAS Number: 80945-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR