Itopride hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(2-(Dimethylamino)ethoxy)benzyl)-3,4-dimethoxybenzamide hydrochloride, N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4-dimethoxybenzamide hydrochloride, topride HCl

A953061-100G A953061-100MG A953061-1G A953061-250MG A953061-25G A953061-5G A953061-1MG
77118-976EA 157.59 USD
77118-976 77118-978 77118-980 77118-982 77118-984 77118-986 77686-978
Itopride hydrochloride 98%
Itopride hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₆N₂O₄·HCl
MW: 394.90 g/mol
MDL Number: MFCD00881710
CAS Number: 122892-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR