3,5-Dichloro-4-fluorobenzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dichloro-2-fluoro-5-(trifluoromethyl)benzene, 3,5-Dichloro-4-fluorobenzotrifluoride

A950331-1G A950331-25G A950331-5G
77118-970EA 41.38 USD
77118-970 77118-972 77118-974
3,5-Dichloro-4-fluorobenzotrifluoride 98%
3,5-Dichloro-4-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₄
MW: 232.99 g/mol
MDL Number: MFCD00068183
CAS Number: 77227-81-7
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR