(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-2-Amino-1,1-diphenyl-1-propanol, 1,1-Diphenyl-L-alaninol, (S)-2-Amino-1,1-diphenylpropan-1-ol

A946438-10G A946438-1G A946438-250MG A946438-5G
77118-962EA 271.6 USD
77118-962 77118-964 77118-966 77118-968
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO
MW: 227.302 g/mol
MDL Number: MFCD00145253
CAS Number: 78603-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR