4-Chloro-2,6-difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A941955-1G A941955-250MG A941955-5G
77118-944EA 82.4 USD
77118-944 77118-946 77118-948
4-Chloro-2,6-difluorophenylboronic acid 98%
4-Chloro-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
MDL Number: MFCD03095138
CAS Number: 925911-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR