Benzyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate 95%

Supplier: Ambeed
Warning

A938992-1G A938992-250MG A938992-5G
77118-934EA 114.58 USD
77118-934 77118-936 77118-938
Benzyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate 95%
Benzyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃
MW: 231.24 g/mol
MDL Number: MFCD11111856
CAS Number: 99027-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR