4-Isobutylpiperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A932274-100G A932274-25G A932274-500G
77118-916EA 52.53 USD
77118-916 77118-918 77118-920
4-Isobutylpiperidine-2,6-dione 98%
4-Isobutylpiperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₉H₁₅NO₂
MDL Number: MFCD18277013
CAS Number: 916982-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR