4-Bromo-2,6-bis(trifluoromethyl)quinoline 95%

Supplier: Ambeed
Warning

A924719-1G A924719-250MG
77118-894EA 37.07 USD
77118-894 77118-896
4-Bromo-2,6-bis(trifluoromethyl)quinoline 95%
4-Bromo-2,6-bis(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₁H₄BrF₆N
MDL Number: MFCD12197304
CAS Number: 35853-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR