[1,1'-Biphenyl]-3,3'-diol 95%

Supplier: Ambeed
Danger

A924040-100MG A924040-1G
77118-892EA 27.59 USD
77118-892 77686-958
[1,1'-Biphenyl]-3,3'-diol 95%
[1,1'-Biphenyl]-3,3'-diol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.20 g/mol
MDL Number: MFCD00094710
CAS Number: 612-76-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR