4-Methyl-5-nitropicolinic acid 95%

Supplier: Ambeed
Warning

A922815-100MG A922815-1G A922815-250MG A922815-5G
77118-884EA 68.96 USD
77118-884 77118-886 77118-888 77118-890
4-Methyl-5-nitropicolinic acid 95%
4-Methyl-5-nitropicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MW: 182.13 g/mol
MDL Number: MFCD07368386
CAS Number: 5832-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR