(4-Nitrophenyl)methanesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-Nitro-α-toluenesulphonyl chloride

A922747-10G A922747-1G A922747-250MG A922747-25G A922747-5G
77118-874EA 59.42 USD
77118-874 77118-876 77118-878 77118-880 77118-882
(4-Nitrophenyl)methanesulfonyl chloride 95%
(4-Nitrophenyl)methanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄S
MW: 235.64 g/mol
MDL Number: MFCD00034030
CAS Number: 4025-75-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR