1-(2-Bromoethoxy)-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A916257-100G A916257-1G A916257-25G A916257-5G
77118-858EA 305.54 USD
77118-858 77118-860 77118-862 77118-864
1-(2-Bromoethoxy)-4-nitrobenzene 95%
1-(2-Bromoethoxy)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
MDL Number: MFCD00031363
CAS Number: 13288-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR