5-Hydroxydecanoate sodium salt 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxydecanoate sodium, Sodium 5-hydroxydecanoate

A913675-100MG A913675-1G A913675-250MG A913675-50MG
77118-838EA 40.18 USD
77118-838 77118-840 77118-842 77118-844
5-Hydroxydecanoate sodium salt 98%
5-Hydroxydecanoate sodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₉O₃·Na
MDL Number: MFCD00153808
CAS Number: 71186-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR