Pivaloylacetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile

A913304-25G A913304-500G
77118-834EA 6.37 USD
77118-834 77118-836
Pivaloylacetonitrile 98%
Pivaloylacetonitrile

Precaución: for Research Use Only

Formula: C₇H₁₁NO
MW: 125.17 g/mol
MDL Number: MFCD00010208
CAS Number: 59997-51-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR