5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone 97%

Supplier: Ambeed
Warning

Synonyms: 5-Ethyl-3-hydroxy-4-methylfuran-2(5H)-one, Ethyl Fenugreek Lactone

A911941-100G A911941-25G A911941-5G
77118-828EA 245.07 USD
77118-828 77118-830 77118-832
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone 97%
5-Ethyl-3-hydroxy-4-methyl-2(5H)-furanone

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.15 g/mol
MDL Number: MFCD00036673
CAS Number: 698-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR