2,7-Dichloroquinoline-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dichloroquinoline-3-carbaldehyde

A907524-100MG A907524-1G A907524-250MG
77118-806EA 43.5 USD
77118-806 77118-808 77118-810
2,7-Dichloroquinoline-3-carboxaldehyde 95%
2,7-Dichloroquinoline-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₅Cl₂NO
MW: 226.059 g/mol
MDL Number: MFCD06202348
CAS Number: 73568-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR