tert-Butyl 9H-pyrido[3,4-b]indole-3-carboxylate 95%

Supplier: Ambeed
Warning

A906406-100MG A906406-250MG
77118-802EA 153.84 USD
77118-802 77118-804
tert-Butyl 9H-pyrido[3,4-b]indole-3-carboxylate 95%
tert-Butyl 9H-pyrido[3,4-b]indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₂
MW: 268.31 g/mol
MDL Number: MFCD06407911
CAS Number: 93835-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR