3-Nitrobenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: m-Nitrobenzonitrile

A903423-100G A903423-25G A903423-500G A903423-5G
77118-776EA 63.66 USD
77118-776 77118-778 77118-780 77118-782
3-Nitrobenzonitrile 98%
3-Nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
MDL Number: MFCD00007194
CAS Number: 619-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR