2-Methylpyrazine 98%

Supplier: Ambeed
Danger

A902038-100G A902038-10G A902038-25G
77118-770EA 12.74 USD
77118-770 77118-772 77118-774
2-Methylpyrazine 98%
2-Methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₆N₂
MW: 94.12 g/mol
MDL Number: MFCD00006142
CAS Number: 109-08-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR