(2-Chloro-5-fluoropyridin-4-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoro-4-(hydroxymethyl)pyridine

A893121-1G A893121-250MG
77118-748EA 31.93 USD
77118-748 77118-750
(2-Chloro-5-fluoropyridin-4-yl)methanol 97%
(2-Chloro-5-fluoropyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.5600 g/mol
MDL Number: MFCD05662385
CAS Number: 884494-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR