(4-Amino-2-nitrophenyl)methanol 97%

Supplier: Ambeed
Warning

A886848-100MG A886848-1G A886848-250MG A886848-5G
77118-722EA 44.28 USD
77118-722 77118-724 77118-726 77118-728
(4-Amino-2-nitrophenyl)methanol 97%
(4-Amino-2-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD01664412
CAS Number: 22996-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR