4-Methylisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 7-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

A881909-1G A881909-5G
77118-718EA 48.81 USD
77118-718 77118-720
4-Methylisatoic anhydride 97%
4-Methylisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD03426400
CAS Number: 63480-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR