(4-Bromo-3-fluorophenyl)methanamine 96%

Supplier: Ambeed
Warning

A881131-100MG A881131-1G A881131-250MG A881131-5G
77118-710EA 29.71 USD
77118-710 77118-712 77118-714 77118-716
(4-Bromo-3-fluorophenyl)methanamine 96%
(4-Bromo-3-fluorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.03 g/mol
MDL Number: MFCD12546114
CAS Number: 581812-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR