3-(Hydroxymethyl)indazole 97%

Supplier: Ambeed
Warning

Synonyms: (1H-Indazol-3-yl)methanol

A879970-1G A879970-250MG A879970-5G
77118-706EA 31.83 USD
77118-706 77118-708 77686-919
3-(Hydroxymethyl)indazole 97%
3-(Hydroxymethyl)indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD05260509
CAS Number: 64132-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR