SB-742457 98+%

Supplier: Ambeed
Warning

Synonyms: Intepirdine, 3-(Phenylsulphonyl)-8-(piperazin-1-yl)quinoline

A879799-10MG A879799-1MG A879799-25MG A879799-5MG
77118-698EA 47.75 USD
77118-698 77118-700 77118-702 77118-704
SB-742457 98+%
SB-742457

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₃O₂S
MW: 353.44 g/mol
MDL Number: MFCD12828872
CAS Number: 607742-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR