Gallic acid monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

A876809-1000G A876809-100G A876809-10G A876809-25G A876809-500G A876809-5G
77118-678EA 71.09 USD
77118-678 77118-680 77118-682 77118-684 77118-686 77118-688
Gallic acid monohydrate 98%
Gallic acid monohydrate

Precaución: for Research Use Only

Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
MDL Number: MFCD00149098
CAS Number: 5995-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR