2-(Methylamino)pyridine 98%

Supplier: Ambeed
Warning

A876344-1G A876344-25G A876344-5G
77118-672EA 11.67 USD
77118-672 77118-674 77118-676
2-(Methylamino)pyridine 98%
2-(Methylamino)pyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00006250
CAS Number: 4597-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR