2-Bromo-5-chloro-1,3-difluorobenzene 98%

Supplier: Ambeed
Warning

A875552-100G A875552-1G A875552-25G
77118-668EA 586.68 USD
77118-668 77118-670 77595-414
2-Bromo-5-chloro-1,3-difluorobenzene 98%
2-Bromo-5-chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
MDL Number: MFCD06798002
CAS Number: 883546-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR