2-(2-Bromophenyl)naphthalene 98%

Supplier: Ambeed
Warning

A874554-250MG A874554-1G
77118-666EA 11.33 USD
77118-666 77686-913
2-(2-Bromophenyl)naphthalene 98%
2-(2-Bromophenyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₆H₁₁Br
MDL Number: MFCD18415513
CAS Number: 22082-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR