9-Propyl-9H-carbazole 95%

Supplier: Ambeed
Warning

A871663-1000G A871663-100G A871663-10G A871663-25G A871663-500G A871663-5G
77118-654EA 204.76 USD
77118-654 77118-656 77118-658 77118-660 77118-662 77118-664
9-Propyl-9H-carbazole 95%
9-Propyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
MDL Number: MFCD00218284
CAS Number: 1484-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR