2,4,5-Trifluorobenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,4,5-trifluorobenzoylchloride

A869083-1G A869083-25G A869083-5G
77118-646EA 10.61 USD
77118-646 77118-648 77118-650
2,4,5-Trifluorobenzoyl chloride 98%
2,4,5-Trifluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
MDL Number: MFCD00061204
CAS Number: 88419-56-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR