4-Bromo-2-hydroxybenzamide 95%

Supplier: Ambeed
Warning

A866671-100MG A866671-250MG A866671-1G
77118-622EA 12.36 USD
77118-622 77118-624 77552-634
4-Bromo-2-hydroxybenzamide 95%
4-Bromo-2-hydroxybenzamide

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD07780674
CAS Number: 5428-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR