3-Bromo[1,1'-biphenyl]-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-[1,1'-biphenyl]-4-ol

A866455-1G A866455-250MG
77118-618EA 39.26 USD
77118-618 77118-620
3-Bromo[1,1'-biphenyl]-4-ol 98%
3-Bromo[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
MDL Number: MFCD00053297
CAS Number: 92-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR