4-Chloro-5-nitrobenzene-1,2-diamine 95%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-5-nitro-o-phenylenediamine

A865705-100MG A865705-1G A865705-250MG
77118-612EA 31.83 USD
77118-612 77118-614 77118-616
4-Chloro-5-nitrobenzene-1,2-diamine 95%
4-Chloro-5-nitrobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
MW: 187.583 g/mol
MDL Number: MFCD03427594
CAS Number: 67073-39-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR