3-Bromo-4-hydroxyquinoline 98+%

Supplier: Ambeed
Danger

Synonyms: 3-Bromoquinolin-4-ol

A862323-1G A862323-250MG A862323-25G A862323-5G
77118-596EA 115.64 USD
77118-596 77118-598 77118-600 77118-602
3-Bromo-4-hydroxyquinoline 98+%
3-Bromo-4-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.054 g/mol
MDL Number: MFCD00234483
CAS Number: 64965-47-5
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR