tert-Butyl 2-(1,3-dioxoisoindolin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A861767-100MG A861767-1G A861767-250MG
77118-590EA 5.16 USD
77118-590 77118-592 77118-594
tert-Butyl 2-(1,3-dioxoisoindolin-2-yl)acetate 95%
tert-Butyl 2-(1,3-dioxoisoindolin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD00023082
CAS Number: 6297-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR