2-Methyl-4-(piperazin-1-yl)quinoline 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Methylquinolin-4-yl)piperazine

A860507-100MG A860507-1G A860507-250MG A860507-5G
77118-580EA 47.75 USD
77118-580 77118-582 77118-584 77118-586
2-Methyl-4-(piperazin-1-yl)quinoline 97%
2-Methyl-4-(piperazin-1-yl)quinoline

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃
MW: 227.305 g/mol
MDL Number: MFCD01935745
CAS Number: 82241-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR