3-Bromophenylboronic acid MIDA ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

A859108-10G A859108-1G A859108-5G
77118-572EA 214.31 USD
77118-572 77118-574 77118-576
3-Bromophenylboronic acid MIDA ester 95%
3-Bromophenylboronic acid MIDA ester

Precaución: for Research Use Only

Formula: C₁₁H₁₁BBrNO₄
MW: 311.92 g/mol
MDL Number: MFCD11215220
CAS Number: 943552-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR