4,5-Dihydro-1H-imidazole-2-sulfonic acid 98%

Supplier: Ambeed
Warning

A856898-100MG A856898-1G A856898-250MG A856898-5G
77118-564EA 16.48 USD
77118-564 77118-566 77118-568 77118-570
4,5-Dihydro-1H-imidazole-2-sulfonic acid 98%
4,5-Dihydro-1H-imidazole-2-sulfonic acid

Precaución: for Research Use Only

Formula: C₃H₆N₂O₃S
MDL Number: MFCD03426388
CAS Number: 64205-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR