Monosodium urate 95%

Supplier: Ambeed
Warning

A855239-1G A855239-250MG
77118-560EA 18.04 USD
77118-560 77118-562
Monosodium urate 95%
Monosodium urate

Precaución: for Research Use Only

Formula: C₅H₃N₄O₃·Na
MDL Number: MFCD00067310
CAS Number: 1198-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR